Benzyl Morpholinoacetate
Dodavatel: Apollo Scientific
OR470085-25G
OR470085-5G
APOSOR470085-25GEA
15970
CZK
APOSOR470085-25G
APOSOR470085-5G
Benzyl Morpholinoacetate
Benzyl Morpholinoacetate
Vzorec:
C₁₃H₁₇NO₃ MW: 235.279 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16495872 CAS číslo: 53342-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...