Tisk…

N-Boc-Pro-Gly-OMe

Dodavatel: Apollo Scientific

OR470084-1G OR470084-5G
APOSOR470084-1GEA 7280 CZK
APOSOR470084-1G APOSOR470084-5G
N-Boc-Pro-Gly-OMe
N-Boc-Pro-Gly-OMe
Vzorec: C₁₃H₂₂N₂O₅
MW: 286,324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16495870
CAS číslo: 34290-72-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR