4-Chloro-2-pyrrolidinobenzoic acid
Dodavatel: Apollo Scientific
OR470068-1G
OR470068-5G
APOSOR470068-1GEA
7280
CZK
APOSOR470068-1G
APOSOR470068-5G
4-Chloro-2-pyrrolidinobenzoic acid
4-Chloro-2-pyrrolidinobenzoic acid
Vzorec:
C₁₁H₁₂ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13191618 CAS číslo: 1197193-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...