Tisk…

3-Bromo-4-(4-methyl-1-piperazinyl)benzoic acid

Dodavatel: Apollo Scientific

OR470064-1G OR470064-5G
APOSOR470064-1GEA 7280 CZK
APOSOR470064-1G APOSOR470064-5G
3-Bromo-4-(4-methyl-1-piperazinyl)benzoic acid
3-Bromo-4-(4-methyl-1-piperazinyl)benzoic acid
Vzorec: C₁₂H₁₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11110915
CAS číslo: 1131622-55-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR