Tisk…

8-Hydroxy-5,6,7,8-tetrahydroquinoline hydrochloride

Dodavatel: Apollo Scientific

OR470051-25G OR470051-5G
APOSOR470051-25GEA 23480 CZK
APOSOR470051-25G APOSOR470051-5G
8-Hydroxy-5,6,7,8-tetrahydroquinoline hydrochloride
8-Hydroxy-5,6,7,8-tetrahydroquinoline hydrochloride
Vzorec: C₉H₁₁NO·ClH
MW: 185,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828697
CAS číslo: 1186663-25-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR