Tisk…

2-(2-Hydroxyethylamino)-4'-nitroacetanilide

Dodavatel: Apollo Scientific

OR470043-25G OR470043-5G
APOSOR470043-25GEA 20300 CZK
APOSOR470043-25G APOSOR470043-5G
2-(2-Hydroxyethylamino)-4'-nitroacetanilide
2-(2-Hydroxyethylamino)-4'-nitroacetanilide
Vzorec: C₁₀H₁₃N₃O₄
MW: 239,228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198896
CAS číslo: 925920-70-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR