Tisk…

3-Bromo-4-(1-imidazolyl)benzoic acid

Dodavatel: Apollo Scientific

OR470021-25G OR470021-5G
APOSOR470021-25GEA 22920 CZK
APOSOR470021-25G APOSOR470021-5G
3-Bromo-4-(1-imidazolyl)benzoic acid
3-Bromo-4-(1-imidazolyl)benzoic acid
Vzorec: C₁₀H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11656807
CAS číslo: 1141669-53-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR