3-(Dimethylamino)-1-(4-methoxyphenyl)propan-1-one
Dodavatel: Apollo Scientific
OR470018-1G
OR470018-5G
APOSOR470018-1GEA
4370
CZK
APOSOR470018-1G
APOSOR470018-5G
3-(Dimethylamino)-1-(4-methoxyphenyl)propan-1-one
3-(Dimethylamino)-1-(4-methoxyphenyl)propan-1-one
Vzorec:
C₁₂H₁₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00454125 CAS číslo: 2138-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...