Tisk…

N-Ethyl-4-aminobenzylamine

Dodavatel: Apollo Scientific

OR470014-1G OR470014-5G
APOSOR470014-1GEA 4370 CZK
APOSOR470014-1G APOSOR470014-5G
N-Ethyl-4-aminobenzylamine
N-Ethyl-4-aminobenzylamine
Vzorec: C₉H₁₄N₂
MW: 150,221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09965833
CAS číslo: 1019115-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR