Tisk…

4-(Bromomethyl)furan-2(5H)-one

Dodavatel: Apollo Scientific
Nebezpečí

OR470013-1G OR470013-5G
APOSOR470013-1GEA 7840 CZK
APOSOR470013-1G APOSOR470013-5G
4-(Bromomethyl)furan-2(5H)-one
4-(Bromomethyl)furan-2(5H)-one
Vzorec: C₅H₅BrO₂
MW: 176,996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052345
CAS číslo: 61934-55-2
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR