N-(5-Bromopyrazin-2-yl)-2-amino-4-methoxyaniline
Dodavatel: Apollo Scientific
OR470002-1G
OR470002-250MG
APOSOR470002-1GEA
17130
CZK
APOSOR470002-1G
APOSOR470002-250MG
N-(5-Bromopyrazin-2-yl)-2-amino-4-methoxyaniline
N-(5-Bromopyrazin-2-yl)-2-amino-4-methoxyaniline
Vzorec:
C₁₁H₁₁BrN₄O Storage Temperature: Ambient |
MDL Number:
MFCD09998891 CAS číslo: 950845-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...