Tisk…

N-(5-Methylsulphonyl-2-nitrophenyl)piperazine

Dodavatel: Apollo Scientific

OR470001-1G OR470001-5G
APOSOR470001-1GEA 9000 CZK
APOSOR470001-1G APOSOR470001-5G
N-(5-Methylsulphonyl-2-nitrophenyl)piperazine
N-(5-Methylsulphonyl-2-nitrophenyl)piperazine
Vzorec: C₁₁H₁₅N₃O₄S
MW: 285,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10565655
CAS číslo: 1095010-42-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR