Tisk…

(3-tert-Butyl-4-pyrrolidin-1-ylphenyl)boronic acid 95%

Dodavatel: Apollo Scientific

OR46773-1G OR46773-250MG
APOSOR46773-1GEA 18620 CZK
APOSOR46773-1G APOSOR46773-250MG
(3-tert-Butyl-4-pyrrolidin-1-ylphenyl)boronic acid 95%
(3-tert-Butyl-4-pyrrolidin-1-ylphenyl)boronic acid
Vzorec: C₁₄H₂₂BNO₂
MW: 247,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28167356
CAS číslo: 895543-00-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR