1-Methylbicyclo[3.1.0]hexan-4-one, technická kvalita
Dodavatel: Apollo Scientific
OR46772-1G
OR46772-250MG
APOSOR46772-1GEA
13230
CZK
APOSOR46772-1G
APOSOR46772-250MG
1-Methylbicyclo[3.1.0]hexan-4-one, technická kvalita
1-Methylbicyclo[3.1.0]hexan-4-one
Vzorec:
C₇H₁₀O MW: 110,154 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24689027 CAS číslo: 14845-46-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...