Tisk…

2-Amino-7-bromoindole hydrochloride

Dodavatel: Apollo Scientific

OR46770-1G OR46770-250MG
APOSOR46770-1GEA 18620 CZK
APOSOR46770-1G APOSOR46770-250MG
2-Amino-7-bromoindole hydrochloride
2-Amino-7-bromoindole hydrochloride
Vzorec: C₈H₇BrN₂·ClH
MW: 247,52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640235
CAS číslo: 2416262-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR