Tisk…

(4-Bromo-3-methylisoxazol-5-yl)methyl acetate 97%

Dodavatel: Apollo Scientific
Varování

OR46746-1G OR46746-250MG
APOSOR46746-1GEA 5720 CZK
APOSOR46746-1G APOSOR46746-250MG
(4-Bromo-3-methylisoxazol-5-yl)methyl acetate 97%
(4-Bromo-3-methylisoxazol-5-yl)methyl acetate
Vzorec: C₇H₈BrNO₃
MW: 234,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24387374
CAS číslo: 1380089-33-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR