Tisk…

1-(3,5-Dichloro-2-hydroxy-4,6-dimethylphenyl)ethanone

Dodavatel: Apollo Scientific
Varování

OR46745-1G OR46745-250MG
APOSOR46745-1GEA 4470 CZK
APOSOR46745-1G APOSOR46745-250MG
1-(3,5-Dichloro-2-hydroxy-4,6-dimethylphenyl)ethanone
1-(3,5-Dichloro-2-hydroxy-4,6-dimethylphenyl)ethanone
Vzorec: C₁₀H₁₀Cl₂O₂
MW: 233.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066842
CAS číslo: 501659-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR