Tisk…

N-(1,3-Benzothiazol-2-yl)benzenesulphonamide

Dodavatel: Apollo Scientific

OR46734-1G OR46734-250MG OR46734-500MG
APOSOR46734-1GEA 18820 CZK
APOSOR46734-1G APOSOR46734-250MG APOSOR46734-500MG
N-(1,3-Benzothiazol-2-yl)benzenesulphonamide
N-(1,3-Benzothiazol-2-yl)benzenesulphonamide
Vzorec: C₁₃H₁₀N₂O₂S₂
MW: 290,361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00432916
CAS číslo: 13068-60-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR