Tisk…

2-(2-Thienylmethylene)quinuclidin-3-one

Dodavatel: Apollo Scientific

OR46712-1G OR46712-100MG OR46712-250MG OR46712-500MG
APOSOR46712-1GEA 37030 CZK
APOSOR46712-1G APOSOR46712-100MG APOSOR46712-250MG APOSOR46712-500MG
2-(2-Thienylmethylene)quinuclidin-3-one
2-(2-Thienylmethylene)quinuclidin-3-one
Vzorec: C₁₂H₁₃NOS
MW: 219,303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00111528
CAS číslo: 82955-33-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR