Tisk…

2-Formylbenzoyl chloride

Dodavatel: Apollo Scientific

OR46684-1G OR46684-250MG OR46684-500MG OR46684-2.5G
APOSOR46684-1GEA 19350 CZK
APOSOR46684-1G APOSOR46684-250MG APOSOR46684-500MG APOSOR46684-2.5G
2-Formylbenzoyl chloride
2-Formylbenzoyl chloride
Vzorec: C₈H₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD13173767
CAS číslo: 117886-88-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR