Tisk…

2-Amino-6-chlorobenzoyl chloride

Dodavatel: Apollo Scientific

OR46669-1G OR46669-50MG OR46669-250MG OR46669-500MG OR46669-5G
APOSOR46669-1GEA 14150 CZK
APOSOR46669-1G APOSOR46669-50MG APOSOR46669-250MG APOSOR46669-500MG APOSOR46669-5G
2-Amino-6-chlorobenzoyl chloride
2-Amino-6-chlorobenzoyl chloride
Vzorec: C₇H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD13174154
CAS číslo: 227328-16-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR