2-Oxo-2,3-dihydro-1H-indole-5-carbaldehyde
Dodavatel: Apollo Scientific
OR46636-1G
OR46636-100MG
OR46636-250MG
OR46636-500MG
APOSOR46636-1GEA
66790
CZK
APOSOR46636-1G
APOSOR46636-100MG
APOSOR46636-250MG
APOSOR46636-500MG
2-Oxo-2,3-dihydro-1H-indole-5-carbaldehyde
2-Oxo-2,3-dihydro-1H-indole-5-carbaldehyde
Vzorec:
C₉H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD02179604 CAS číslo: 612487-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...