Tisk…

1-Ethyl-1H-imidazol-5-amine

Dodavatel: Apollo Scientific

OR46627-1G OR46627-100MG OR46627-250MG OR46627-500MG
APOSOR46627-1GEA 69100 CZK
APOSOR46627-1G APOSOR46627-100MG APOSOR46627-250MG APOSOR46627-500MG
1-Ethyl-1H-imidazol-5-amine
1-Ethyl-1H-imidazol-5-amine
Vzorec: C₅H₉N₃
MW: 111,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19205887
CAS číslo: 99979-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR