Tisk…

2-Bromo-3-chlorobenzenesulphonamide

Dodavatel: Apollo Scientific
Varování

OR46616-1G OR46616-250MG OR46616-5G
APOSOR46616-1GEA 2650 CZK
APOSOR46616-1G APOSOR46616-250MG APOSOR46616-5G
2-Bromo-3-chlorobenzenesulphonamide
2-Bromo-3-chlorobenzenesulphonamide
Vzorec: C₆H₅BrClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD18390303
CAS číslo: 1261499-12-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR