Ethyl 5-bromothiophene-3-carboxylate 97+%
Dodavatel: Apollo Scientific
|
Varování
|
OR46588-5G
OR46588-1G
APOSOR46588-5GEA
2320
CZK
APOSOR46588-5G
APOSOR46588-1G
Ethyl 5-bromothiophene-3-carboxylate 97+%
Ethyl 5-bromothiophene-3-carboxylate
Vzorec:
C₇H₇BrO₂S MW: 235,098 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14155650 CAS číslo: 170355-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...