Tisk…

2-(Methylsulphonyl)-10H-phenothiazine

Dodavatel: Apollo Scientific
Varování

OR46581-1G OR46581-5G
APOSOR46581-1GEA 8700 CZK
APOSOR46581-1G APOSOR46581-5G
2-(Methylsulphonyl)-10H-phenothiazine
2-(Methylsulphonyl)-10H-phenothiazine
Vzorec: C₁₃H₁₁NO₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD20926232
CAS číslo: 23503-68-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR