Tisk…

4-Bromo-1H-benzimidazole-6-carbonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-6-cyano-1H-benzimidazole, 4-Bromo-1H-benzo[d]imidazole-6-carbonitrile

OR46541-1G OR46541-250MG OR46541-5G
APOSOR46541-1GEA 3110 CZK
APOSOR46541-1G APOSOR46541-250MG APOSOR46541-5G
4-Bromo-1H-benzimidazole-6-carbonitrile
4-Bromo-1H-benzimidazole-6-carbonitrile
Vzorec: C₈H₄BrN₃
MW: 222,041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22558167
CAS číslo: 1360921-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR