Tisk…

3-Bromo-4,5-diaminobenzonitrile 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-5-cyanobenzene-1,2-diamine, 3-Bromo-5-cyanophenylene-1,2-diamine

OR46540-1G OR46540-250MG OR46540-5G
APOSOR46540-1GEA 5890 CZK
APOSOR46540-1G APOSOR46540-250MG APOSOR46540-5G
3-Bromo-4,5-diaminobenzonitrile 95%
3-Bromo-4,5-diaminobenzonitrile
Vzorec: C₇H₆BrN₃
MW: 212,047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628913
CAS číslo: 1417371-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR