Tisk…

3-Bromo-5-iodobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Bromo-5-iodophenyl)methanol

OR46537-1G OR46537-250MG
APOSOR46537-1GEA 4140 CZK
APOSOR46537-1G APOSOR46537-250MG
3-Bromo-5-iodobenzyl alcohol
3-Bromo-5-iodobenzyl alcohol
Vzorec: C₇H₆BrIO
MW: 312,93 g/mol
Bod tání: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD09745849
CAS číslo: 188813-08-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR