4-Amino-2-(methylthio)pyrimidine-5-carboxamide 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR4632-1G
OR4632-250MG
OR4632-5G
APOSOR4632-1GEA
5000
CZK
APOSOR4632-1G
APOSOR4632-250MG
APOSOR4632-5G
4-Amino-2-(methylthio)pyrimidine-5-carboxamide 95+%
4-Amino-2-(methylthio)pyrimidine-5-carboxamide
Vzorec:
C₆H₈N₄OS Storage Temperature: Ambient |
MDL Number:
MFCD00126685 CAS číslo: 89533-28-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...