2-(Bromomethyl)pyrazine hydrobromide 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR46239-1G
OR46239-5G
APOSOR46239-1GEA
5820
CZK
APOSOR46239-1G
APOSOR46239-5G
2-(Bromomethyl)pyrazine hydrobromide 95%
2-(Bromomethyl)pyrazine hydrobromide
Vzorec:
C₅H₅BrN₂·BrH Storage Temperature: Ambient |
MDL Number:
MFCD23105761 CAS číslo: 1421789-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...