Dihydrolevoglucosenone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(1S,5R)-4-Oxo-6,8-dioxabicyclo[3.2.1]octane, (1S,5R)-6,8-Dioxabicyclo[3.2.1]octan-4-one, Cyrene™
OR46234-1G
OR46234-100G
APOSOR46234-1GEA
622
CZK
APOSOR46234-1G
APOSOR46234-100G
Dihydrolevoglucosenone
Dihydrolevoglucosenone
Vzorec:
C₆H₈O₃ MW: 128,126 g/mol Bod varu: 227 °C Bod tání: < –20 °C Hustota: 1,25 g/cm³ (25 °C) Bod vzplanutí: 108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD23105760 CAS číslo: 53716-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...