Tisk…

3-(Chlorosulphonyl)azetidine, N-BOC protected

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: tert-Butyl 3-(chlorosulphonyl)azetidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-3-(chlorosulphonyl)azetidine

OR46226-1G OR46226-250MG
APOSOR46226-1GEA 16140 CZK
APOSOR46226-1G APOSOR46226-250MG
3-(Chlorosulphonyl)azetidine, N-BOC protected
3-(Chlorosulphonyl)azetidine, N-BOC protected
Vzorec: C₈H₁₄ClNO₄S
Storage Temperature: Freezer
MDL Number: MFCD11040529
CAS číslo: 1310732-18-3
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR