5,7-Dibromoisoquinoline 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR46139-1G
OR46139-250MG
APOSOR46139-1GEA
27780
CZK
APOSOR46139-1G
APOSOR46139-250MG
5,7-Dibromoisoquinoline 95%
5,7-Dibromoisoquinoline
Vzorec:
C₉H₅Br₂N MW: 286,951 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31457100 CAS číslo: 2137673-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...