Tisk…

5-Bromo-2-iodo-1H-indole 95%

Dodavatel: Apollo Scientific
Varování

OR46137-1G OR46137-250MG
APOSOR46137-1GEA 31840 CZK
APOSOR46137-1G APOSOR46137-250MG
5-Bromo-2-iodo-1H-indole 95%
5-Bromo-2-iodo-1H-indole
Vzorec: C₈H₅BrIN
MW: 321,94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27987772
CAS číslo: 1572177-90-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR