5-Bromo-6-azaindole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-1H-pyrrolo[2,3-c]pyridine
OR46136-5G
APOSOR46136-5GEA
3610
CZK
APOSOR46136-5G
5-Bromo-6-azaindole 98%
5-Bromo-6-azaindole
Vzorec:
C₇H₅BrN₂ MW: 197,032 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12024306 CAS číslo: 1215387-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...