5-Bromo-1H-imidazo[4,5-b]pyridine, 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-3H-imidazo[4,5-b]pyridine, 5-Bromo-4-azabenzimidazole, 5-Bromo-1H-imidazo[4,5-b]pyridine
OR46119-1G
OR46119-5G
APOSOR46119-1GEA
3150
CZK
APOSOR46119-1G
APOSOR46119-5G
5-Bromo-1H-imidazo[4,5-b]pyridine, 97%
5-Bromo-1H-imidazo[4,5-b]pyridine,
Vzorec:
C₆H₄BrN₃ Storage Temperature: Ambient |
MDL Number:
MFCD11227145 CAS číslo: 28279-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...