3-Bromo-5-nitrophenol 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromo-5-hydroxynitrobenzene
OR46064-10G
OR46064-1G
OR46064-5G
APOSOR46064-10GEA
8300
CZK
APOSOR46064-10G
APOSOR46064-1G
APOSOR46064-5G
3-Bromo-5-nitrophenol 96%
3-Bromo-5-nitrophenol
Vzorec:
C₆H₄BrNO₃ MW: 218,005 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09965960 CAS číslo: 116632-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...