2,6-Dimethylnicotinaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
OR46031-1G
OR46031-5G
APOSOR46031-1GEA
6480
CZK
APOSOR46031-1G
APOSOR46031-5G
2,6-Dimethylnicotinaldehyde
2,6-Dimethylnicotinaldehyde
Vzorec:
C₈H₉NO Hustota: 1,07 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03265780 CAS číslo: 650141-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...