Tisk…

2-Bromo-6-hydroxybenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Bromo-2-cyanophenol

OR46022-1G OR46022-5G
APOSOR46022-1GEA 662 CZK
APOSOR46022-1G APOSOR46022-5G
2-Bromo-6-hydroxybenzonitrile
2-Bromo-6-hydroxybenzonitrile
Vzorec: C₇H₄BrNO
MW: 198.017 g/mol
Bod tání: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD12828440
CAS číslo: 73289-85-7
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR