1,1,1-Trichloro-3-iodopropane 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR460057-1G
OR460057-250MG
APOSOR460057-1GEA
17460
CZK
APOSOR460057-1G
APOSOR460057-250MG
1,1,1-Trichloro-3-iodopropane 97%
1,1,1-Trichloro-3-iodopropane
Vzorec:
C₃H₄Cl₃I MW: 273,327 g/mol Bod varu: 110 °C (10 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD31560933 CAS číslo: 23153-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...