Tisk…

1-(Pyridin-3-yl)piperazine dihydrobromide

Dodavatel: Apollo Scientific
Varování

OR460013-250MG
APOSOR460013-250MGEA 12960 CZK
APOSOR460013-250MG
1-(Pyridin-3-yl)piperazine dihydrobromide
1-(Pyridin-3-yl)piperazine dihydrobromide
Vzorec: C₉H₁₃N₃·2BrH
MW: 325,044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11841224
CAS číslo: 1185369-72-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR