Tisk…

1-Benzyl-4-chloropiperidine

Dodavatel: Apollo Scientific
Varování

OR4559-1G OR4559-250MG
APOSOR4559-1GEA 1780 CZK
APOSOR4559-1G APOSOR4559-250MG
1-Benzyl-4-chloropiperidine
1-Benzyl-4-chloropiperidine
Vzorec: C₁₂H₁₆ClN
Storage Temperature: Ambient
MDL Number: MFCD00510091
CAS číslo: 67848-71-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR