Tisk…
2-Amino-1-(4-methyl-1-piperazinyl)-ethanone dihydrochloride 97%
Dodavatel: Apollo Scientific
OR452218-1G OR452218-500MG
APOSOR452218-1GEA 14880 CZK
APOSOR452218-1G APOSOR452218-500MG
2-Amino-1-(4-methyl-1-piperazinyl)-ethanone dihydrochloride 97%
2-Amino-1-(4-methyl-1-piperazinyl)-ethanone dihydrochloride
Vzorec: C₇H₁₅N₃O·2ClH
MW: 230,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796412
CAS číslo: 146788-11-6