1-(4-Methoxyphenyl)-cyclopentanemethanamine 97%
Dodavatel: Apollo Scientific
OR452215-1G
OR452215-250MG
OR452215-100MG
APOSOR452215-1GEA
22980
CZK
APOSOR452215-1G
APOSOR452215-250MG
APOSOR452215-100MG
1-(4-Methoxyphenyl)-cyclopentanemethanamine 97%
1-(4-Methoxyphenyl)-cyclopentanemethanamine
Vzorec:
C₁₃H₁₉NO MW: 205,296 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01817110 CAS číslo: 23528-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...