4-Cyclopropylpyridine-2-carboxylic acid
Dodavatel: Apollo Scientific
OR45221-1G
OR45221-250MG
APOSOR45221-1GEA
23150
CZK
APOSOR45221-1G
APOSOR45221-250MG
4-Cyclopropylpyridine-2-carboxylic acid
4-Cyclopropylpyridine-2-carboxylic acid
Vzorec:
C₉H₉NO₂ MW: 163,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18257015 CAS číslo: 1256807-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...