Tisk…

1-Boc-4-cyano-4-(2-methylphenyl)-piperidine 97%

Dodavatel: Apollo Scientific

OR452205-1G OR452205-250MG OR452205-500MG
APOSOR452205-1GEA 22950 CZK
APOSOR452205-1G APOSOR452205-250MG APOSOR452205-500MG
1-Boc-4-cyano-4-(2-methylphenyl)-piperidine 97%
1-Boc-4-cyano-4-(2-methylphenyl)-piperidine
Vzorec: C₁₈H₂₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11849780
CAS číslo: 186347-28-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR