Tisk…

N-Boc-[1-(aminomethyl)propyl]amine 97%

Dodavatel: Apollo Scientific

OR452201-1G OR452201-250MG OR452201-500MG
APOSOR452201-1GEA 24800 CZK
APOSOR452201-1G APOSOR452201-250MG APOSOR452201-500MG
N-Boc-[1-(aminomethyl)propyl]amine 97%
N-Boc-[1-(aminomethyl)propyl]amine
Vzorec: C₉H₂₀N₂O₂
MW: 188,267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16660180
CAS číslo: 732219-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR