N-Boc-[(1S)-1-(aminomethyl)propyl]amine 97%
Dodavatel: Apollo Scientific
OR452200-1G
OR452200-250MG
OR452200-500MG
APOSOR452200-1GEA
24800
CZK
APOSOR452200-1G
APOSOR452200-250MG
APOSOR452200-500MG
N-Boc-[(1S)-1-(aminomethyl)propyl]amine 97%
N-Boc-[(1S)-1-(aminomethyl)propyl]amine
Vzorec:
C₉H₂₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18831342 CAS číslo: 251325-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...