(S)-N-Boc-2-aminobutylamine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR452198-1G
OR452198-250MG
OR452198-500MG
APOSOR452198-1GEA
18620
CZK
APOSOR452198-1G
APOSOR452198-250MG
APOSOR452198-500MG
(S)-N-Boc-2-aminobutylamine 97%
(S)-N-Boc-2-aminobutylamine
Vzorec:
C₉H₂₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22397601 CAS číslo: 1336411-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...